Assay Details
Gene Symbol: Tomm40l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083338.1 | NP_001076807.1 | 7 - 8 | 680 | 106 | |
XM_006250247.2 | XP_006250309.1 | 8 - 9 | 1159 | 106 | ||
GenBank mRNA | AB274732.1 | - | 7 - 8 | 680 | 106 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
