Assay Details
Gene Symbol: Ufc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003709.2 | NP_001003709.1 | 5 - 6 | 634 | 91 | |
XM_006250251.2 | XP_006250313.1 | 6 - 7 | 589 | 91 | ||
GenBank mRNA | AB186051.1 | - | 5 - 6 | 552 | 91 | |
AB441169.1 | - | 5 - 6 | 581 | 91 | ||
BC087648.1 | - | 5 - 6 | 634 | 91 | ||
FQ212748.1 | - | 5 - 6 | 652 | 91 | ||
FQ213023.1 | - | 5 - 6 | 625 | 91 | ||
FQ213507.1 | - | 5 - 6 | 635 | 91 | ||
FQ216057.1 | - | 5 - 6 | 629 | 91 | ||
FQ218926.1 | - | 5 - 6 | 655 | 91 | ||
FQ219875.1 | - | 5 - 6 | 504 | 91 | ||
FQ232457.1 | - | 5 - 6 | 611 | 91 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
