Assay Details
Gene Symbol: Ppox | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105968.1 | NP_001099438.1 | 2 - 3 | 396 | 105 | |
XM_006250240.2 | XP_006250302.1 | 1 - 2 | 1264 | 105 | ||
XM_006250241.1 | XP_006250303.1 | 2 - 3 | 428 | 105 | ||
XM_006250243.1 | XP_006250305.1 | 1 - 2 | 88 | 105 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the XM_006250243.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
