Assay Details
Gene Symbol: Traf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008769908.1 | XP_008768130.1 | 3 - 4 | 660 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
