Assay Details
Gene Symbol: Sdhc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005534.1 | NP_001005534.1 | 2 - 3 | 88 | 69 | |
XM_008769731.1 | XP_008767953.1 | 3 - 4 | 135 | 69 | ||
GenBank mRNA | BC082027.1 | - | 2 - 3 | 88 | 69 | |
FQ210665.1 | - | 2 - 3 | 90 | 69 | ||
FQ213406.1 | - | 2 - 3 | 107 | 69 | ||
FQ215668.1 | - | 3 - 4 | 142 | 69 | ||
FQ215712.1 | - | 2 - 3 | 90 | 69 | ||
FQ216122.1 | - | 2 - 3 | 106 | 69 | ||
FQ216612.1 | - | 2 - 3 | 107 | 69 | ||
FQ216658.1 | - | 2 - 3 | 90 | 69 | ||
FQ216892.1 | - | 2 - 3 | 106 | 69 | ||
FQ217679.1 | - | 2 - 3 | 107 | 69 | ||
FQ224704.1 | - | 2 - 3 | 90 | 69 | ||
FQ225195.1 | - | 2 - 3 | 101 | 69 | ||
FQ229651.1 | - | 2 - 3 | 107 | 69 | ||
FQ231805.1 | - | 2 - 3 | 107 | 69 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the XM_008769731.1 transcripts.
This assay is in the 5'UTR region of the XM_008769731.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
