Assay Details
Gene Symbol: Cadm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047103.1 | NP_001040568.1 | 5 - 6 | 692 | 91 | |
XM_006250323.2 | XP_006250385.1 | 6 - 7 | 964 | 91 | ||
GenBank mRNA | BC161811.1 | - | 5 - 6 | 693 | 91 | |
DQ272743.1 | - | 5 - 6 | 692 | 91 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
