Assay Details
Gene Symbol: Igsf8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014787.1 | NP_001014787.1 | 7 - 8 | 1921 | 96 | |
XM_006250286.2 | XP_006250348.1 | 7 - 8 | 2080 | 96 | ||
XM_006250288.2 | XP_006250350.1 | 6 - 7 | 2022 | 96 | ||
XM_006250289.2 | XP_006250351.1 | 6 - 7 | 1859 | 96 | ||
GenBank mRNA | BC092192.1 | - | 7 - 8 | 1921 | 96 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
