Assay Details
Gene Symbol: Pcmtd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107810.1 | NP_001101280.2 | 6 - 7 | 993 | 97 | |
GenBank mRNA | FQ211587.1 | - | 6 - 7 | 992 | 97 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P009678 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 705197 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 508291 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 134362 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 741966 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 494329 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 315634 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 12125 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 360162 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 319526 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 295309 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 338561 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 711703 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 404076 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 70521 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 101306 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 98767 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P009678 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P009678 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P009678 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P501623 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 4748 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 52498 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10843131 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10843128 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10843127 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10843129 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10843130 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10843125 | - | - |
Rat Genome 230 2.0 Array | 1374454_at | - | - |
Rat Genome 230 2.0 Array | 1374454_PM_at | - | - |
Rat Genome 230 2.0 Array | 1375916_PM_at | - | - |
Rat Genome 230 2.0 Array | 1375916_at | - | - |
Affymetrix HT Rat Focus Array | 1374454_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1375916_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1374454_at | - | - |
Affymetrix HT Rat Focus Array | 1375916_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0300019408.rn.1 | TC0300002613.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0300019409.rn.1 | TC0300002613.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0300019411.rn.1 | TC0300002613.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0300019410.rn.1 | TC0300002613.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0300019412.rn.1 | TC0300002613.rn.1 | RTA TC |
Rat Expression Set 230 | 1375916_PM_at | - | - |
Rat Expression Set 230 | 1374454_PM_at | - | - |
Rat Expression Set 230 | 1374454_at | - | - |
Rat Expression Set 230 | 1375916_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10843125 | - | - |
Rat Genome U34 Set | rc_AI639429_f_at | - | - |
Rat Genome U34 Set | rc_AI639155_at | - | - |
Rat Genome U34 Set | rc_AA955163_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21705310 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1374830 | - | - |
