Assay Details
Gene Symbol: Zgpat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009656.1 | NP_001009656.1 | 4 - 5 | 1109 | 79 | |
XM_006235749.1 | XP_006235811.1 | 5 - 6 | 1105 | 79 | ||
XM_006235750.1 | XP_006235812.1 | 5 - 6 | 1070 | 79 | ||
XM_006235751.2 | XP_006235813.1 | 5 - 6 | 1091 | 79 | ||
XM_006235752.2 | XP_006235814.1 | 4 - 5 | 1005 | 79 | ||
GenBank mRNA | BC087720.1 | - | 4 - 5 | 1109 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
