Assay Details
Gene Symbol: Rac1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134366.1 | NP_599193.1 | 5 - 6 | 451 | 94 | |
XM_008768986.1 | XP_008767208.1 | 6 - 7 | 542 | 94 | ||
GenBank mRNA | AY491395.1 | - | 5 - 6 | 451 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of protein phosphorylation
auditory receptor cell morphogenesis
mast cell chemotaxis
epithelial cell morphogenesis
endocytosis
phagocytosis, engulfment
chemotaxis
hyperosmotic response
cytoskeleton organization
actin filament organization
cell adhesion
G protein-coupled receptor signaling pathway
small GTPase mediated signal transduction
axon guidance
metabolic process
cell population proliferation
positive regulation of lamellipodium assembly
positive regulation of cell-substrate adhesion
regulation of neuron maturation
dendrite development
cell migration
Rac protein signal transduction
cerebral cortex radially oriented cell migration
embryonic olfactory bulb interneuron precursor migration
cerebral cortex GABAergic interneuron development
regulation of cell morphogenesis
lamellipodium assembly
actin cytoskeleton organization
actin filament polymerization
regulation of cell migration
positive regulation of actin filament polymerization
ruffle organization
negative regulation of interleukin-23 production
substrate adhesion-dependent cell spreading
positive regulation of phosphatidylinositol 3-kinase activity
engulfment of apoptotic cell
cell-cell junction organization
bone resorption
positive regulation of DNA replication
anatomical structure arrangement
dendrite morphogenesis
cell motility
homeostasis of number of cells within a tissue
positive regulation of stress fiber assembly
localization within membrane
positive regulation of focal adhesion assembly
synaptic transmission, GABAergic
Wnt signaling pathway, planar cell polarity pathway
regulation of respiratory burst
cellular response to mechanical stimulus
semaphorin-plexin signaling pathway
dopaminergic neuron differentiation
protein localization to plasma membrane
positive regulation of neutrophil chemotaxis
cochlea morphogenesis
ruffle assembly
positive regulation of substrate adhesion-dependent cell spreading
GTPase activity
protein binding
GTP binding
enzyme binding
protein kinase binding
GTP-dependent protein binding
thioesterase binding
histone deacetylase binding
Rho GDP-dissociation inhibitor binding
positive regulation of protein phosphorylation
auditory receptor cell morphogenesis
mast cell chemotaxis
epithelial cell morphogenesis
endocytosis
phagocytosis, engulfment
chemotaxis
hyperosmotic response
cytoskeleton organization
actin filament organization
cell adhesion
G protein-coupled receptor signaling pathway
small GTPase mediated signal transduction
axon guidance
metabolic process
cell population proliferation
positive regulation of lamellipodium assembly
positive regulation of cell-substrate adhesion
regulation of neuron maturation
dendrite development
cell migration
Rac protein signal transduction
cerebral cortex radially oriented cell migration
embryonic olfactory bulb interneuron precursor migration
cerebral cortex GABAergic interneuron development
regulation of cell morphogenesis
lamellipodium assembly
actin cytoskeleton organization
actin filament polymerization
regulation of cell migration
positive regulation of actin filament polymerization
ruffle organization
negative regulation of interleukin-23 production
substrate adhesion-dependent cell spreading
positive regulation of phosphatidylinositol 3-kinase activity
engulfment of apoptotic cell
cell-cell junction organization
bone resorption
positive regulation of DNA replication
anatomical structure arrangement
dendrite morphogenesis
cell motility
homeostasis of number of cells within a tissue
positive regulation of stress fiber assembly
localization within membrane
positive regulation of focal adhesion assembly
synaptic transmission, GABAergic
Wnt signaling pathway, planar cell polarity pathway
regulation of respiratory burst
cellular response to mechanical stimulus
semaphorin-plexin signaling pathway
dopaminergic neuron differentiation
protein localization to plasma membrane
positive regulation of neutrophil chemotaxis
cochlea morphogenesis
ruffle assembly
positive regulation of substrate adhesion-dependent cell spreading
GTPase activity
protein binding
GTP binding
enzyme binding
protein kinase binding
GTP-dependent protein binding
thioesterase binding
histone deacetylase binding
Rho GDP-dissociation inhibitor binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P395709 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 615772 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 627309 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 763281 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 491330 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 171955 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 923185 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 755423 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 664442 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 946516 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 254357 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 767585 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 315834 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 226672 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 276821 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 175131 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P395709 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P395709 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P395709 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P162583 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P395709 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 36439 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 67908 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10756726 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10756727 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10756723 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10756722 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10756728 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10756724 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10756721 | - | - |
Rat Genome 230 2.0 Array | 1372513_at | - | - |
Rat Genome 230 2.0 Array | 1388332_at | - | - |
Rat Genome 230 2.0 Array | 1372513_PM_at | - | - |
Rat Genome 230 2.0 Array | 1391048_PM_at | - | - |
Rat Genome 230 2.0 Array | 1388332_PM_at | - | - |
Rat Genome 230 2.0 Array | 1391048_at | - | - |
Affymetrix HT Rat Focus Array | 1372513_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1372513_at | - | - |
Affymetrix HT Rat Focus Array | 1391048_at | - | - |
Affymetrix HT Rat Focus Array | 1391048_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1200001603.rn.1 | TC1200000223.rn.1 | RTA TC |
Rat Expression Set 230 | 1388332_PM_at | - | - |
Rat Expression Set 230 | 1372513_at | - | - |
Rat Expression Set 230 | 1372513_PM_at | - | - |
Rat Expression Set 230 | 1388332_at | - | - |
Rat Expression Set 230 | 1391048_at | - | - |
Rat Expression Set 230 | 1391048_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10756721 | - | - |
Rat Genome U34 Set | rc_AA799542_at | - | - |
Rat Genome U34 Set | rc_AI103616_at | - | - |
Rat Genome U34 Set | AA848238_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21885804 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1355225 | - | - |
RatRef-12 Expression BeadChip | ILMN_56272 | - | - |
