Assay Details
Gene Symbol: Rac1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008768986.1 | XP_008767208.1 | 3 - 4 | 262 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
