Assay Details
Gene Symbol: Snx8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105912.1 | NP_001099382.1 | 9 - 10 | 1155 | 112 | |
XM_006248920.2 | XP_006248982.1 | 9 - 10 | 1138 | 112 | ||
XM_006248921.2 | XP_006248983.1 | 9 - 10 | 1538 | 112 | ||
XM_006248923.2 | XP_006248985.1 | 9 - 10 | 1411 | 112 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P1056309 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 352825 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 624369 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 316980 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 50752 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 940370 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 343209 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 946914 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 910131 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 959942 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 441755 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 730645 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 4321 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 283136 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 749557 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 356994 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 952852 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 435168 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 701805 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 622456 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 69353 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 390071 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 427242 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 44689 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 122854 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 494033 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 147707 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 936057 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 451109 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 168756 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 957008 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 347344 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 423295 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 921816 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 127808 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 125786 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 126890 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 45896 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 171112 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 180157 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 99463 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P1056309 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P1056309 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P1056309 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P1056309 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 41372 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 81187 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10756964 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10756960 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10756958 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10756955 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10756961 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10756963 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10756962 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10756954 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10756956 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10756957 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10756959 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10756953 | - | - |
Rat Genome 230 2.0 Array | 1380802_at | - | - |
Rat Genome 230 2.0 Array | 1383751_PM_at | - | - |
Rat Genome 230 2.0 Array | 1383751_at | - | - |
Rat Genome 230 2.0 Array | 1380802_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1380802_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1380802_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1200002092.rn.1 | TC1200000282.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1200002091.rn.1 | TC1200000282.rn.1 | RTA TC |
Rat Expression Set 230 | 1380802_PM_at | - | - |
Rat Expression Set 230 | 1383751_at | - | - |
Rat Expression Set 230 | 1383751_PM_at | - | - |
Rat Expression Set 230 | 1380802_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10756953 | - | - |
Rat Genome U34 Set | rc_AI070058_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21524572 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1352389 | - | - |
