Assay Details
Gene Symbol: Pgm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106007.1 | NP_001099477.1 | 9 - 10 | 1252 | 71 | |
XM_006250996.2 | XP_006251058.1 | 10 - 11 | 1355 | 71 | ||
XM_006250997.2 | XP_006251059.1 | 11 - 12 | 1462 | 71 | ||
GenBank mRNA | BC160893.1 | - | 10 - 11 | 1352 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
