Assay Details
Gene Symbol: Gabra4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080587.3 | NP_542154.3 | 6 - 7 | 814 | 117 | |
XM_008770135.1 | XP_008768357.1 | 5 - 6 | 1501 | 117 | ||
GenBank mRNA | S55933.1 | - | 6 - 7 | 814 | 117 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
