Assay Details
Gene Symbol: Srd5a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013990.1 | NP_001014012.1 | 4 - 5 | 604 | 77 | |
XM_006250897.1 | XP_006250959.1 | 4 - 5 | 642 | 77 | ||
GenBank mRNA | BC086584.1 | - | 4 - 5 | 604 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
