Assay Details
Gene Symbol: Rell1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113776.1 | NP_001107248.1 | 4 - 5 | 515 | 162 | |
XM_006250998.2 | XP_006251060.1 | 4 - 5 | 610 | 162 | ||
GenBank mRNA | BC158872.1 | - | 4 - 5 | 515 | 162 | |
FQ224412.1 | - | 4 - 5 | 442 | 162 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
