Assay Details
Gene Symbol: Rell1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006250998.2 | XP_006251060.1 | 6 - 7 | 995 | 91 | |
GenBank mRNA | FQ224412.1 | - | 6 - 7 | 827 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
