Assay Details
Gene Symbol: Nsun7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017452.1 | NP_001017452.1 | 4 - 5 | 798 | 86 | |
XM_006250946.2 | XP_006251008.1 | 4 - 5 | 1089 | 86 | ||
XM_006250947.2 | XP_006251009.1 | 4 - 5 | 1092 | 86 | ||
XM_006250948.2 | XP_006251010.1 | 4 - 5 | 1089 | 86 | ||
GenBank mRNA | BC083653.1 | - | 4 - 5 | 798 | 86 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
