Assay Details
Gene Symbol: Noa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006959.1 | NP_001006960.1 | 2 - 3 | 1393 | 116 | |
XM_006250845.2 | XP_006250907.1 | 2 - 3 | 1708 | 116 | ||
GenBank mRNA | BC083902.1 | - | 2 - 3 | 1393 | 116 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
