Assay Details
Gene Symbol: Noa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006250845.2 | XP_006250907.1 | 5 - 6 | 2163 | 128 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
