Assay Details
Gene Symbol: Fip1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008295.1 | NP_001008296.1 | 13 - 14 | 1233 | 60 | |
XM_006250882.2 | XP_006250944.1 | 16 - 17 | 1541 | 60 | ||
XM_006250883.2 | XP_006250945.1 | 15 - 16 | 1514 | 60 | ||
XM_006250884.2 | XP_006250946.1 | 15 - 16 | 1470 | 60 | ||
XM_006250885.2 | XP_006250947.1 | 15 - 16 | 1431 | 60 | ||
XM_006250887.2 | XP_006250949.1 | 14 - 15 | 1360 | 60 | ||
GenBank mRNA | BC085767.1 | - | 13 - 14 | 1233 | 60 | |
EU224276.1 | - | 13 - 14 | 1232 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P154565 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 277603 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 896109 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 675500 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 826988 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 876306 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 913797 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 609370 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 488291 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 381334 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 623676 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 833132 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 89862 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 737072 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 438614 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 371521 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 520517 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 107187 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 857412 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 558677 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 207899 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 554826 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 828536 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 198319 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 339786 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 667734 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 834470 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 491921 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 343453 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 74504 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 246394 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 233518 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 496071 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 582069 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 872648 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 717332 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 844287 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 936697 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 515257 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 893913 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 823084 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 300950 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 228355 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 135665 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 695580 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 464065 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 357084 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 580551 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 17503 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 7939 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 119851 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 93043 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 19093 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 32551 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P154565 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P154565 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P154565 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P496788 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 92383 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 95126 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 82521 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 42699 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 99108 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10776495 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10776500 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10776489 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10776504 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10776491 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10776508 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10776503 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10776498 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10776492 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10776488 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10776507 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10776502 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10776493 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10776490 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10776501 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10776509 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10776496 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10776505 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10776487 | - | - |
Rat Genome 230 2.0 Array | 1389621_PM_at | - | - |
Rat Genome 230 2.0 Array | 1389621_at | - | - |
Affymetrix HT Rat Focus Array | 1389621_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1389621_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1400012342.rn.1 | TC1400001673.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1400012345.rn.1 | TC1400001673.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1400012341.rn.1 | TC1400001673.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1400012343.rn.1 | TC1400001673.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1400012344.rn.1 | TC1400001673.rn.1 | RTA TC |
Rat Expression Set 230 | 1389621_at | - | - |
Rat Expression Set 230 | 1389621_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10776487 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 22172728 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1355940 | - | - |
RatRef-12 Expression BeadChip | ILMN_65978 | - | - |
