Assay Details
Gene Symbol: Cenpc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004098.1 | NP_001004098.1 | 16 - 17 | 2444 | 146 | |
GenBank mRNA | AY693786.1 | - | 9 - 10 | 1287 | 146 | |
AY693788.1 | - | 16 - 17 | 2444 | 146 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
