Assay Details
Gene Symbol: Cenpc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004098.1 | NP_001004098.1 | 18 - 19 | 2689 | 104 | |
GenBank mRNA | AY693786.1 | - | 11 - 12 | 1532 | 104 | |
AY693788.1 | - | 18 - 19 | 2689 | 104 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P466023 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 183971 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 228492 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 966901 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 296253 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 771346 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 319820 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 853776 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 421299 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 447393 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 100852 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 47772 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 805377 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 409425 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 319753 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 359435 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 191984 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 500201 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 90624 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 377976 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 558024 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 591086 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 912596 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 266229 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 722674 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 887075 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 609983 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 929555 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 364688 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 504184 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 82953 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 270870 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 889121 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 869398 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 852279 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 329850 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 672987 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 453599 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 358195 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 488964 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 104145 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 151697 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 175471 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 35405 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 37710 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 157351 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 176550 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 173070 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P466023 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P466023 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P466023 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P466023 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P18977 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 71263 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 38680 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 11148 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 96994 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10772117 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10772121 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10772125 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10772111 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10772120 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10772112 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10772115 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10772122 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10772119 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10772113 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10772123 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10772110 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10772114 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10772124 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10772109 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10772116 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10772118 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10772108 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10772107 | - | - |
Affymetrix Rat Clariom S Assay | PSR1400002758.rn.1 | TC1400000330.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1400002760.rn.1 | TC1400000330.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1400002757.rn.1 | TC1400000330.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1400002759.rn.1 | TC1400000330.rn.1 | RTA TC |
Affymetrix Rat Gene 1.0 ST Array | 10772107 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21410890 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1376601 | - | - |
