Assay Details
Gene Symbol: Cenpc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004098.1 | NP_001004098.1 | 7 - 8 | 804 | 141 | |
GenBank mRNA | AY693787.1 | - | 2 - 3 | 228 | 141 | |
AY693788.1 | - | 7 - 8 | 804 | 141 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
