Assay Details
Gene Symbol: Cenpc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004098.1 | NP_001004098.1 | 8 - 9 | 1391 | 143 | |
GenBank mRNA | AY693786.1 | - | 1 - 2 | 234 | 143 | |
AY693787.1 | - | 3 - 4 | 815 | 143 | ||
AY693788.1 | - | 8 - 9 | 1391 | 143 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
