Assay Details
Gene Symbol: Cenpc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004098.1 | NP_001004098.1 | 9 - 10 | 1568 | 151 | |
GenBank mRNA | AY693786.1 | - | 2 - 3 | 411 | 151 | |
AY693787.1 | - | 4 - 5 | 992 | 151 | ||
AY693788.1 | - | 9 - 10 | 1568 | 151 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
