Assay Details
Gene Symbol: Usp46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191596.1 | NP_001178525.1 | 8 - 9 | 1069 | 157 | |
XM_006250888.2 | XP_006250950.1 | 8 - 9 | 1579 | 157 | ||
XM_006250889.2 | XP_006250951.1 | 8 - 9 | 1632 | 157 | ||
XM_008770137.1 | XP_008768359.1 | 8 - 9 | 1662 | 157 | ||
GenBank mRNA | FQ211578.1 | - | 8 - 9 | 1117 | 157 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
