Assay Details
Gene Symbol: Ociad2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271181.1 | NP_001258110.1 | 2 - 3 | 160 | 106 | |
GenBank mRNA | FM129783.1 | - | 3 - 4 | 175 | 106 | |
FM139638.1 | - | 2 - 3 | 160 | 106 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
