Assay Details
Gene Symbol: Uchl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017237.3 | NP_058933.2 | 1 - 2 | 80 | 58 | |
GenBank mRNA | BC060573.1 | - | 1 - 2 | 84 | 58 | |
D10699.1 | - | 1 - 2 | 64 | 58 | ||
FQ214229.1 | - | 1 - 2 | 76 | 58 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
