Assay Details
Gene Symbol: G3bp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013989.1 | NP_001014011.1 | 3 - 4 | 550 | 139 | |
XM_006250733.2 | XP_006250795.1 | 3 - 4 | 544 | 139 | ||
XM_006250734.2 | XP_006250796.1 | 3 - 4 | 599 | 139 | ||
XM_006250735.1 | XP_006250797.1 | 3 - 4 | 382 | 139 | ||
XM_006250736.2 | XP_006250798.1 | 2 - 3 | 193 | 139 | ||
GenBank mRNA | BC079225.1 | - | 3 - 4 | 550 | 139 | |
FQ212261.1 | - | 3 - 4 | 387 | 139 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
