Assay Details
Gene Symbol: Dck | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024158.1 | NP_077072.1 | 5 - 6 | 688 | 61 | |
GenBank mRNA | FQ212366.1 | - | 993 | 61 | ||
L33894.1 | - | 5 - 6 | 688 | 61 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
