Assay Details
Gene Symbol: Areg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017123.1 | NP_058819.1 | 1 - 2 | 236 | 131 | |
GenBank mRNA | X55183.1 | - | 1 - 2 | 236 | 131 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
