Assay Details
Gene Symbol: Ap5m1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030036.1 | NP_001025207.1 | 1 - 2 | 301 | 71 | |
GenBank mRNA | BC099829.1 | - | 1 - 2 | 301 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
