Assay Details
Gene Symbol: Ap5m1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030036.1 | NP_001025207.1 | 4 - 5 | 1315 | 182 | |
GenBank mRNA | BC099829.1 | - | 4 - 5 | 1315 | 182 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
