Assay Details
Gene Symbol: Rab2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037645.1 | NP_001032734.1 | 4 - 5 | 369 | 73 | |
XM_006251883.2 | XP_006251945.1 | 5 - 6 | 469 | 73 | ||
GenBank mRNA | BC107443.1 | - | 4 - 5 | 369 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
