Assay Details
Gene Symbol: Hnrnpc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277600.1 | NP_001264529.1 | 3 - 4 | 449 | 83 | |
NM_001277601.1 | NP_001264530.1 | 3 - 4 | 449 | 83 | ||
NM_001277602.1 | NP_001264531.1 | 2 - 3 | 423 | 83 | ||
XM_006251858.2 | XP_006251920.1 | 2 - 3 | 917 | 83 | ||
XM_006251859.2 | XP_006251921.1 | 2 - 3 | 410 | 83 | ||
XM_006251861.2 | XP_006251923.1 | 2 - 3 | 916 | 83 | ||
XM_006251862.2 | XP_006251924.1 | 4 - 5 | 600 | 83 | ||
XM_006251863.2 | XP_006251925.1 | 2 - 3 | 410 | 83 | ||
XM_006251864.2 | XP_006251926.1 | 4 - 5 | 505 | 83 | ||
XM_006251865.2 | XP_006251927.1 | 2 - 3 | 916 | 83 | ||
XM_006251866.1 | XP_006251928.1 | 3 - 4 | 436 | 83 | ||
XM_008770508.1 | XP_008768730.1 | 3 - 4 | 944 | 83 | ||
XM_008770509.1 | XP_008768731.1 | 3 - 4 | 2012 | 83 | ||
XM_008770510.1 | XP_008768732.1 | 3 - 4 | 444 | 83 | ||
XM_008770511.1 | XP_008768733.1 | 3 - 4 | 444 | 83 | ||
XM_008770512.1 | XP_008768734.1 | 3 - 4 | 436 | 83 | ||
GenBank mRNA | BC097346.1 | - | 2 - 3 | 373 | 83 | |
CB582273.1 | - | 3 - 4 | 399 | 83 | ||
FQ214011.1 | - | 1 - 2 | 60 | 83 | ||
FQ222540.1 | - | 1 - 2 | 99 | 83 | ||
FQ227266.1 | - | 1 - 2 | 99 | 83 |
Gene Symbol: LOC100911576 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271241.1 | NP_001258170.1 | 3 - 4 | 414 | 83 | |
XM_003751394.2 | XP_003751442.1 | 3 - 4 | 400 | 83 | ||
XM_003751395.2 | XP_003751443.1 | 3 - 4 | 400 | 83 | ||
XM_006251449.2 | XP_006251511.1 | 3 - 4 | 2011 | 83 | ||
XM_006251450.2 | XP_006251512.1 | 2 - 3 | 915 | 83 | ||
XM_006251451.1 | XP_006251513.1 | 2 - 3 | 374 | 83 | ||
XM_006251452.2 | XP_006251514.1 | 3 - 4 | 400 | 83 | ||
XM_006251453.1 | XP_006251515.1 | 4 - 5 | 564 | 83 | ||
XM_006251454.1 | XP_006251516.1 | 4 - 5 | 469 | 83 | ||
XM_006251455.2 | XP_006251517.1 | 2 - 3 | 914 | 83 | ||
XM_006251456.1 | XP_006251518.1 | 2 - 3 | 374 | 83 | ||
XM_006251457.1 | XP_006251519.1 | 2 - 3 | 374 | 83 | ||
XM_006251458.2 | XP_006251520.1 | 3 - 4 | 400 | 83 | ||
XM_008770242.1 | XP_008768464.1 | 3 - 4 | 908 | 83 | ||
XM_008770243.1 | XP_008768465.1 | 3 - 4 | 400 | 83 | ||
GenBank mRNA | AY724499.1 | - | 2 - 3 | 842 | 83 | |
FQ214198.1 | - | 1 - 2 | 99 | 83 | ||
FQ215781.1 | - | 1 - 2 | 99 | 83 | ||
FQ227893.1 | - | 1 - 2 | 99 | 83 | ||
FQ228082.1 | - | 1 - 2 | 99 | 83 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
