Assay Details
Gene Symbol: Hnrnpc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277600.1 | NP_001264529.1 | 7 - 8 | 901 | 113 | |
NM_001277601.1 | NP_001264530.1 | 7 - 8 | 862 | 113 | ||
XM_006251858.2 | XP_006251920.1 | 6 - 7 | 1369 | 113 | ||
XM_006251859.2 | XP_006251921.1 | 6 - 7 | 862 | 113 | ||
XM_006251861.2 | XP_006251923.1 | 6 - 7 | 1329 | 113 | ||
XM_006251862.2 | XP_006251924.1 | 8 - 9 | 1013 | 113 | ||
XM_006251863.2 | XP_006251925.1 | 6 - 7 | 823 | 113 | ||
XM_006251864.2 | XP_006251926.1 | 8 - 9 | 918 | 113 | ||
XM_008770508.1 | XP_008768730.1 | 7 - 8 | 1396 | 113 | ||
XM_008770509.1 | XP_008768731.1 | 7 - 8 | 2464 | 113 | ||
GenBank mRNA | BC097346.1 | - | 6 - 7 | 780 | 113 |
Gene Symbol: LOC100911576 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271241.1 | NP_001258170.1 | 7 - 8 | 827 | 113 | |
XM_003751395.2 | XP_003751443.1 | 7 - 8 | 852 | 113 | ||
XM_006251449.2 | XP_006251511.1 | 7 - 8 | 2463 | 113 | ||
XM_006251450.2 | XP_006251512.1 | 6 - 7 | 1367 | 113 | ||
XM_006251451.1 | XP_006251513.1 | 6 - 7 | 826 | 113 | ||
XM_006251453.1 | XP_006251515.1 | 8 - 9 | 977 | 113 | ||
XM_006251454.1 | XP_006251516.1 | 8 - 9 | 882 | 113 | ||
XM_006251455.2 | XP_006251517.1 | 6 - 7 | 1327 | 113 | ||
XM_006251456.1 | XP_006251518.1 | 6 - 7 | 787 | 113 | ||
XM_008770242.1 | XP_008768464.1 | 7 - 8 | 1360 | 113 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
