Assay Details
Gene Symbol: Hnrnpc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277601.1 | NP_001264530.1 | 4 - 5 | 530 | 69 | |
NM_001277602.1 | NP_001264531.1 | 3 - 4 | 504 | 69 | ||
XM_006251861.2 | XP_006251923.1 | 3 - 4 | 997 | 69 | ||
XM_006251862.2 | XP_006251924.1 | 5 - 6 | 681 | 69 | ||
XM_006251863.2 | XP_006251925.1 | 3 - 4 | 491 | 69 | ||
XM_006251864.2 | XP_006251926.1 | 5 - 6 | 586 | 69 | ||
XM_006251865.2 | XP_006251927.1 | 3 - 4 | 997 | 69 | ||
XM_006251866.1 | XP_006251928.1 | 4 - 5 | 517 | 69 | ||
XM_008770512.1 | XP_008768734.1 | 4 - 5 | 517 | 69 | ||
GenBank mRNA | BC097346.1 | - | 3 - 4 | 454 | 69 | |
FQ214011.1 | - | 2 - 3 | 141 | 69 | ||
FQ222540.1 | - | 2 - 3 | 180 | 69 | ||
FQ227266.1 | - | 2 - 3 | 180 | 69 |
Gene Symbol: LOC100911576 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271241.1 | NP_001258170.1 | 4 - 5 | 495 | 69 | |
XM_003751394.2 | XP_003751442.1 | 4 - 5 | 481 | 69 | ||
XM_006251453.1 | XP_006251515.1 | 5 - 6 | 645 | 69 | ||
XM_006251454.1 | XP_006251516.1 | 5 - 6 | 550 | 69 | ||
XM_006251455.2 | XP_006251517.1 | 3 - 4 | 995 | 69 | ||
XM_006251456.1 | XP_006251518.1 | 3 - 4 | 455 | 69 | ||
XM_006251457.1 | XP_006251519.1 | 3 - 4 | 455 | 69 | ||
XM_006251458.2 | XP_006251520.1 | 4 - 5 | 481 | 69 | ||
GenBank mRNA | AY724499.1 | - | 3 - 4 | 923 | 69 | |
FQ214198.1 | - | 2 - 3 | 180 | 69 | ||
FQ227893.1 | - | 2 - 3 | 180 | 69 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
