Assay Details
Gene Symbol: Psmc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100509.1 | NP_001093979.1 | 6 - 7 | 488 | 87 | |
GenBank mRNA | BC107950.1 | - | 6 - 7 | 409 | 87 | |
DY570284.1 | - | 6 - 7 | 488 | 87 | ||
FQ213949.1 | - | 6 - 7 | 436 | 87 | ||
FQ214043.1 | - | 6 - 7 | 441 | 87 | ||
FQ227742.1 | - | 6 - 7 | 445 | 87 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
