Assay Details
Gene Symbol: Psmc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100509.1 | NP_001093979.1 | 9 - 10 | 770 | 130 | |
GenBank mRNA | BC107950.1 | - | 9 - 10 | 691 | 130 | |
FQ213949.1 | - | 9 - 10 | 718 | 130 | ||
FQ214043.1 | - | 9 - 10 | 723 | 130 | ||
FQ227742.1 | - | 9 - 10 | 727 | 130 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
