Assay Details
Gene Symbol: Tox4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173324.2 | NP_775446.2 | 6 - 7 | 948 | 83 | |
GenBank mRNA | AF267197.1 | - | 5 - 6 | 900 | 83 | |
BC081696.1 | - | 6 - 7 | 948 | 83 | ||
FQ210274.1 | - | 6 - 7 | 956 | 83 | ||
FQ212245.1 | - | 6 - 7 | 956 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
