Assay Details
Gene Symbol: Tox4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173324.2 | NP_775446.2 | 7 - 8 | 1688 | 104 | |
GenBank mRNA | AF267197.1 | - | 6 - 7 | 1640 | 104 | |
BC081696.1 | - | 7 - 8 | 1688 | 104 | ||
FQ210274.1 | - | 7 - 8 | 1696 | 104 | ||
FQ212245.1 | - | 7 - 8 | 1696 | 104 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
