Assay Details
Gene Symbol: Hsd11b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017080.2 | NP_058776.2 | 1 - 2 | 418 | 87 | |
XM_006250471.2 | XP_006250533.1 | 2 - 3 | 199 | 87 | ||
XM_006250473.2 | XP_006250535.1 | 2 - 3 | 234 | 87 | ||
GenBank mRNA | BC078865.1 | - | 1 - 2 | 239 | 87 | |
CK474613.1 | - | 1 - 2 | 435 | 87 | ||
FQ209829.1 | - | 1 - 2 | 185 | 87 | ||
FQ210302.1 | - | 1 - 2 | 183 | 87 | ||
FQ210563.1 | - | 1 - 2 | 183 | 87 | ||
FQ210596.1 | - | 1 - 2 | 185 | 87 | ||
FQ211059.1 | - | 1 - 2 | 182 | 87 | ||
FQ211404.1 | - | 1 - 2 | 184 | 87 | ||
FQ212763.1 | - | 1 - 2 | 186 | 87 | ||
FQ214152.1 | - | 1 - 2 | 185 | 87 | ||
FQ214397.1 | - | 2 - 3 | 241 | 87 | ||
FQ218558.1 | - | 1 - 2 | 183 | 87 | ||
FQ218584.1 | - | 1 - 2 | 185 | 87 | ||
FQ219123.1 | - | 1 - 2 | 185 | 87 | ||
FQ219241.1 | - | 1 - 2 | 184 | 87 | ||
FQ219394.1 | - | 1 - 2 | 183 | 87 | ||
FQ219517.1 | - | 1 - 2 | 185 | 87 | ||
FQ219624.1 | - | 1 - 2 | 118 | 87 | ||
J05107.1 | - | 1 - 2 | 166 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
