Assay Details
Gene Symbol: Usp21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127638.1 | NP_001121110.1 | 2 - 3 | 804 | 65 | |
XM_008769764.1 | XP_008767986.1 | 2 - 3 | 947 | 65 | ||
GenBank mRNA | BC166443.1 | - | 2 - 3 | 804 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
