Assay Details
Gene Symbol: Ephx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034090.3 | NP_001029262.1 | 2 - 3 | 324 | 69 | |
NM_012844.3 | NP_036976.2 | 2 - 3 | 353 | 69 | ||
GenBank mRNA | BC061568.1 | - | 2 - 3 | 284 | 69 | |
M26125.1 | - | 329 | 69 |
Gene Symbol: LOC690147 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008769898.1 | XP_008768120.1 | 3 - 4 | 451 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
