Assay Details
Gene Symbol: Arhgap30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109077.1 | NP_001102547.1 | 10 - 11 | 1504 | 139 | |
GenBank mRNA | FQ211641.1 | - | 1512 | 139 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
