Assay Details
Gene Symbol: Arhgap30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109077.1 | NP_001102547.1 | 5 - 6 | 607 | 101 | |
GenBank mRNA | FQ211641.1 | - | 615 | 101 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
