Assay Details
Gene Symbol: Rin2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107786.1 | NP_001101256.1 | 5 - 6 | 620 | 150 | |
XM_006235142.2 | XP_006235204.1 | 5 - 6 | 2195 | 150 | ||
XM_006235143.2 | XP_006235205.1 | 4 - 5 | 1477 | 150 | ||
XM_006235146.2 | XP_006235208.1 | 4 - 5 | 1147 | 150 | ||
XM_006235147.2 | XP_006235209.1 | 4 - 5 | 1250 | 150 | ||
XM_006235148.2 | XP_006235210.1 | 3 - 4 | 801 | 150 | ||
XM_006235149.2 | XP_006235211.1 | 4 - 5 | 1564 | 150 | ||
XM_006235150.2 | XP_006235212.1 | 3 - 4 | 815 | 150 | ||
XM_006235151.2 | XP_006235213.1 | 2 - 3 | 305 | 150 | ||
XM_008762270.1 | XP_008760492.1 | 6 - 7 | 1638 | 150 | ||
XM_008762271.1 | XP_008760493.1 | 4 - 5 | 1033 | 150 | ||
XM_008762272.1 | XP_008760494.1 | 2 - 3 | 1055 | 150 | ||
XM_008762273.1 | XP_008760495.1 | 2 - 3 | 2076 | 150 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
