Assay Details
Gene Symbol: Rgs19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021661.2 | NP_067693.1 | 2 - 3 | 324 | 59 | |
XM_006235801.2 | XP_006235863.1 | 2 - 3 | 3085 | 59 | ||
GenBank mRNA | AF068136.1 | - | 1 - 2 | 32 | 59 | |
BC088141.1 | - | 2 - 3 | 324 | 59 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
