Assay Details
Gene Symbol: Rgs19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021661.2 | NP_067693.1 | 1 - 2 | 214 | 83 | |
GenBank mRNA | BC088141.1 | - | 1 - 2 | 214 | 83 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC088141.1, NM_021661.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
